Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LDLR Monoclonal Antibody (OTI4E9), TrueMAB™, OriGene
SDP

Catalog No. 501684574
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4574 Supplier OriGene Supplier No. TA812507
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The low density lipoprotein (LDL) receptor system coordinates the metabolism of cholesterol, an essential component of the plasma membrane of all mammalian cells. Study of this system has led to an enhanced understanding of the cellular basis of cholesterol homeostasis. It has also brought into focus an important mechanism of metabolic regulation-the process of receptor-mediated endocytosis (1). Data suggest that the juxtamembranous region of the cytoplasmic domain participates in protein:protein interactions that allow the low density lipoprotein receptor to cluster in coated pits (2). It has been shown that the family of LDL receptors may serve as viral receptors. Endocytosis of the Flaviviridae viruses, hepatitis C virus, GB virus C/hepatitis G virus, and bovine viral diarrheal virus (BVDV) was shown to be mediated by LDL receptors on cultured cells (3).

Specifications

Antigen LDLR
Applications Western Blot
Classification Monoclonal
Clone OTI4E9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene LDLR
Gene Accession No. P01130
Gene Alias FH, FHC, LDLCQ2
Gene Symbols LDLR
Host Species Mouse
Immunogen Full length human recombit protein of human LDLR produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3949
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.