Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LDLR Recombinant Rabbit Monoclonal Antibody (301), APC
GREENER_CHOICE

Catalog No. PIMA540994
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA540994 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA540994 Supplier Invitrogen™ Supplier No. MA540994
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.

The low density lipoprotein receptor (LDL-R) gene family consists of cell surface proteins involved in receptor-mediated endocytosis of specific ligands. Low density lipoprotein (LDL) is normally bound at the cell membrane and taken into the cell ending up in lysosomes where the protein is degraded and the cholesterol is made available for repression of microsomal enzyme 3-hydroxy-3-methylglutaryl coenzyme A (HMG CoA) reductase, the rate-limiting step in cholesterol synthesis. At the same time, a reciprocal stimulation of cholesterol ester synthesis takes place. Mutations in the LDL-R gene cause the autosomal dominant disorder, familial hypercholesterolemia. Along with SCARB1, CLDN1, and the tetraspanin superfamily member CD81, LDL-R has been reported to be an entry factor for the Hepatitis C virus. At least three isoforms of LDL-R are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDLR
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 301
Concentration 0.1 mg/mL
Conjugate APC
Formulation PBS with 0.5% BSA and 0.03% ProClin 300
Gene LDLR
Gene Accession No. P01130
Gene Alias FH; FHC; Hlb301; I79_005860; LDL receptor; LDLA; LDLCQ2; LDLR; ldl-r; LDLRA; low density lipoprotein receptor; low density lipoprotein receptor protein; low-density lipoprotein receptor; low-density lipoprotein receptor class A domain-containing protein 3
Gene Symbols LDLR
Host Species Rabbit
Immunogen Recombinant Human LDL Receptor/LDLR protein.
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3949
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.