Learn More
Invitrogen™ LDLRAD2 Polyclonal Antibody

Description
The antibody detects endogenous levels of total LDLRAD2 protein.
Specifications
Specifications
| Antigen | LDLRAD2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | LDLRAD2 |
| Gene Accession No. | Q5SZI1 |
| Gene Alias | LDLRAD2; low density lipoprotein receptor A domain containing 2; low density lipoprotein receptor class A domain containing 2; low-density lipoprotein receptor class A domain-containing protein 2; lrad2 |
| Gene Symbols | LDLRAD2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.