Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LDN 209929 dihydrochloride

Catalog No. 482810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-281-0 10 mg
48-285-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-281-0 Supplier Tocris Bioscience™ Supplier No. 4828/10
This item is not returnable. View return policy

Potent and selective haspin inhibitor

LDN 209929 dihydrochloride is a potent and selective haspin kinase inhibitor (IC50 = 55 nM). Displays 180-fold selectivity over DYRK2.

Specifications

Purity >98%
CAS 1784281-97-5
Quantity 10 mg
Product Type LDN 209929 dihydrochloride
Molecular Weight (g/mol) 405.77
Inhibitors LDN 209929 dihydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.