Learn More
Aat Bioquest LDS 751 CAS 181885-68-7 (25 mg)
SDP
Supplier:  Aat Bioquest 17561
Encompass_Preferred
LDS 751 is a fluorescent cell-permeant nucleic acid stain that can be well excited with 488 nm laser line although it has a peak excitation at 543 nm on dsDNA. This catalog is for 25 mg.
Catalog No. 50-224-9748