Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Lead
SCI_ED

Catalog No. S25383A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
Ea.
Packaging:
Sheet - 12 x 12 x 0.03 in.
Sheet - 6.5 x 6.5 x 0.03 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25383A Ea. Sheet - 12 x 12 x 0.03 in.
S25383B Ea. Sheet - 6.5 x 6.5 x 0.03 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S25383A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Color Blue-White to Gray
Density 11.3g/mL
Quantity Ea.
Packaging Sheet - 12 x 12 x 0.03 in.
Assay Percent Range 100% w/v
Linear Formula Pb
Formula Weight 207.2
Chemical Name or Material Lead
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.