Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Lead Acetate, Trihydrate
SCI_ED

Catalog No. S25378 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Powder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25378 100 g Powder
S25378A 500 g Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S25378
Add to Cart
Add to Cart
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
IUPAC Name λ²-lead(2+) diacetate trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Color White
Quantity 100 g
Assay Percent Range >95% w/v
Linear Formula Pb(C2H3O2)2·3H2O
Formula Weight 379.33
Packaging Powder
Density 2.55g/mL at 20°C
Chemical Name or Material Lead Acetate, Trihydrate
Safety handling Check Alert storage code Red
ShelfLife 1 year
DOTInformation DOT Class 6.1, III, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.