Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Acetate, Trihydrate, Crystal, Reagent, ACS, 99-103%, Spectrum™ Chemical
SDP

Catalog No. 18603752 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-752 6 x 500 g Poly Bottle
18-603-750 125 g Poly Bottle
18-603-751 500 g Poly Bottle
18-603-753 2.5 kg Poly Bottle
18-603-754 4 x 2.5 kg Poly Bottle
18-603-755 12 kg Poly Pail
18-603-756 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-603-752 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L1040500GMCS6
Add to Cart
Add to Cart

L1040, 6080-56-4, C4H6O4Pb·3H2O

Spectrum™ Chemical Lead Acetate, Trihydrate, Crystal, Reagent, ACS is a chemical compound and is obtained by treating lead oxide with acetic acid. It is used as a reagent and comes in a crystalline form. As an ACS grade Reagent, Spectrum Chemical manufactured compounds are used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
IUPAC Name λ²-lead(2+) diacetate trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

CAS Max % 100%
Quantity 6 x 500 g
UN Number UN1616
Percent Purity 99 to 103%
Packaging Poly Bottle
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.