Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Acetate TS, 2% (w/v) Alcoholic Solution, Ricca Chemical
SDP

Catalog No. p-3340715 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
42938-16 500 mL Natural Poly Bottle
42938-32 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42938-16 Supplier Ricca Chemical Company Supplier No. 4293.816
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
IUPAC Name λ2-lead(2+) diacetate trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Color Colorless
CAS Min % 2.39, 96.0
CAS Max % 2.49, 100.0
Physical Form Liquid
Quantity 500 mL
Grade Laboratory
Concentration or Composition (by Analyte or Components) 2% (w/v) Alcoholic Solution
Packaging Natural Poly Bottle
Chemical Name or Material Lead Acetate TS, Alcoholic
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.