Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Carbonate, Powder, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. 18603764 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-764 2.5 kg Poly Bottle
18-603-761 125 g Poly Bottle
18-603-762 500 g Poly Bottle
18-603-763 6 x 500 g Poly Bottle
18-603-765 12 kg Poly Pail
18-603-766 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-603-764 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L10452.5KG
Add to Cart
Add to Cart

L1045, 598-63-0, PbCO3

Spectrum™ Chemical Lead Carbonate, Powder, Reagent, ACS is a chemical compound that is created industrially from lead acetate and carbon dioxide. It is an important pigment used in paints, putty, and ceramics. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 598-63-0
Molecular Formula CO3Pb
Molecular Weight (g/mol) 267.20
InChI Key MFEVGQHCNVXMER-UHFFFAOYSA-L
IUPAC Name λ²-lead(2+) carbonate
SMILES [Pb++].[O-]C([O-])=O

Specifications

CAS Max % 100%
Quantity 2.5 kg
Grade ACS
Packaging Poly Bottle
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.