Learn More
Electron Microscopy Sciences Lead Citrate, Trihydrate 25 GR
SDP
Supplier:  Electron Microscopy Sciences 17800
Encompass_Preferred
CAS #512-26-5 Pb(C₆H₅O₇)2·₃H₂O F.W. 1053.82
References: The most widely used metal stain for ultra thin sections. Reynolds, E.S., J. Cell Bio., 17, 208 (1963) Sato, T., J. Electron Micro., 116, 133 (1976) Venable, J.H. and Coggeshall, R. (1965). A simplified lead citrate stain for use in electron microscopy. J. Cell Biol., 25:407 Famy, A. (1967). An extemporaneous lead citrate for electron microscopy. Proc. 25th Ann. EMSA Meet., p.148. Claitor's Pub. Division, Baton Rouge, LA.
Catalog No. 50-268-90
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.