Learn More
Lead(II) bromide, ultra dry, 99.999% (metals basis)
Description
Lead(II) bromide is used in the field of antirust, pigment and photograph. The molten lead(II) bromide acts as an electrolyte. Lead(II) bromide provides a high concentration of lead(II) ions and bromide ions to carry the current during the electrolysis process. The rare-earth-doped alkali-lead bromide crystals (potassium lead bromide (or) KPB; rubidium lead bromide or RPB) emerge as promising new low-phonon-energy host materials for mid-IR applications and are useful for solid state lasers. Hybrid organic/lead halide perovskites are promising materials for solar cell fabrication.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 916°C |
| Melting Point | 373°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99.999% (Metals basis) |
| Solubility Information | Slightly soluble in water. Insoluble in alcohol. Soluble in ammonia and alkali. |
| IUPAC Name | dibromolead |
| Formula Weight | 367.01 |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H302+H332-H360Df-H373 |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P312-P314-P330-P501c |
| DOTInformation | DOT Class: 6.1, Packaging Group: III |
| EINECSNumber | 233-084-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.