Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) chloride, 99%

Catalog No. AC193310010
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC193310010 1 kg Plastic bottle
AC193310500 50 g Plastic bottle
AC193312500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC193310010 Supplier Thermo Scientific Chemicals Supplier No. 193310010
Only null left
Add to Cart

CAS: 7758-95-4 | Cl2Pb | 278.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-95-4
Molecular Formula Cl2Pb
Molecular Weight (g/mol) 278.10
MDL Number MFCD00011157
InChI Key HWSZZLVAJGOAAY-UHFFFAOYSA-L
Synonym lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dry
PubChem CID 24459
SMILES [Cl-].[Cl-].[Pb++]

Specifications

Boiling Point 950.0°C
Melting Point 501.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula PbCl2
Merck Index 15, 5459
Solubility Information Solubility in water: 0.99g/L (20°C). Other solubilities: readly soluble in solution of nh4cl and nh4no3, soluble in alkalis
IUPAC Name λ²-lead(2+) dichloride
Formula Weight 278.11
Percent Purity 99%
Packaging Plastic bottle
Chemical Name or Material Lead(II) chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquatic life with long lasting effects.
May damage the unborn child.
Su
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON C
EINECSNumber 231-845-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.