Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) chloride, Reagent Grade, 99%

Catalog No. AA1234936
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1234936 500 g
AA1234918 50 g
AA12349A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1234936 Supplier Thermo Scientific Chemicals Supplier No. 01234936
Add to Cart

Analytical Reagent, Preparation of Lead Salts, As Solder and Flux | CAS: 7758-95-4 | Cl2Pb | 278.10 g/mol

It is employed in the manufacture of infrared transmitting glass, and ornamental glass called arena glass. It also finds application as analytical reagent, in preparation of lead salts, as solder and flux, for provision of lead titanate, and used in the purification process of ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-95-4
Molecular Formula Cl2Pb
Molecular Weight (g/mol) 278.10
MDL Number MFCD00011157
InChI Key HWSZZLVAJGOAAY-UHFFFAOYSA-L
Synonym lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dry
PubChem CID 24459
IUPAC Name dichlorolead
SMILES [Cl-].[Cl-].[Pb++]

Specifications

Boiling Point 951°C
Melting Point 501°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99%
Solubility Information Soluble in hot water,alkali hydroxides and NH4Cl solution. Insoluble in cold water and alcohol.
Formula Weight 278.1
Grade Reagent
Odor Odorless
Density 5.85
Chemical Name or Material Lead(II) chloride
Show More Show Less
Hazard Category H302+H332-H350-H360Df-H372
Hazard Statement Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 231-845-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.