Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) iodide, ultra dry, 99.999% (metals basis)

Catalog No. AA4431406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4431406 5 g
AA4431414 25 g
AA4431422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4431406 Supplier Thermo Scientific Chemicals Supplier No. 04431406
Add to Cart

Lead(II) Iodide, Ultra Dry, I2Pb, CAS Number-10101-63-0, blei ii-iodid, diiodo-, e2-plumbane, hsdb 636, lead diiodide, lead ii iodide, lead iodide pbi2, pbi2, plumbous iodide, plumbum jodatum, 100g, 99.999% (Metals basis), 954 deg.C, Yellow, 6.16, DOT Class: 6.1, Packaging Group: III, 233-256-9, 461 | CAS: 10101-63-0 | I2Pb | 461.00 g/mol

Lead(II) iodide is used as a detector material for high energy photons including x-rays and gamma rays. It is used in photography, printing, mosaic gold, and bronzing. It exhibits ferroelastic properties and has efficiency in stopping X-ray and gamma ray, which provides excellent environmental stability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-63-0
Molecular Formula I2Pb
Molecular Weight (g/mol) 461.00
MDL Number MFCD00011163
InChI Key RQQRAHKHDFPBMC-UHFFFAOYSA-L
Synonym lead ii iodide, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, diiodo-, e2-plumbane
PubChem CID 24931
IUPAC Name λ²-lead(2+) diiodide
SMILES [I-].[I-].[Pb++]

Specifications

Boiling Point 954°C
Melting Point 402°C
Color Yellow
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Soluble in concentrated solutions of alkali iodides and sodium thiosulfate. Insoluble in alcohol,cold hydrochloric acid and organic solvents.
Formula Weight 461
Odor Odorless
Sensitivity Light sensitive
Density 6.16 g/cm3 at 20°C
Chemical Name or Material Lead(II) iodide, ultra dry
Show More Show Less
Hazard Category H302+H332-H350-H360-H373
Hazard Statement Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P201-P202-P261-P262-P264b-P270-P271-P280-P301+P312-P304+P340-P308+P313-P312-P314-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 233-256-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.