Learn More
Lead(II) metavanadate, 99.9% (metals basis), Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 50 g |
| UN Number | UN2291 |
| Merck Index | 14,5427 |
| Solubility Information | Insoluble in water,decomposes in HNO3 |
| Formula Weight | 405.07 |
| Percent Purity | 99.9% |
| Odor | Odorless |
| Assay | (metals basis) |
| Chemical Name or Material | Lead(II) metavanadate |
Safety and Handling
| Hazard Category | H302+H332-H350-H360Df-H373 |
| Hazard Statement | GHS H Statement H360-H373-H302-H332 May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S. |
| EINECSNumber | 233-248-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.