Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) metavanadate, 99.9% (metals basis), Thermo Scientific™

Catalog No. AA3937830
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3937830 250 g
AA3937818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3937830 Supplier Thermo Scientific Chemicals Supplier No. 3937830

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

As pigment, in manufacturing of vanadium compounds

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-79-3
Molecular Formula O6PbV2
Molecular Weight (g/mol) 405.10
MDL Number MFCD00016271
InChI Key OGRLITDAVSILTM-UHFFFAOYSA-N
Synonym lead vanadate, unii-99565ywk3f, lead ii metavanadate, lead vanadate v, lead divanadium hexaoxide, lead vanadium oxide pbv2o6, 2o3v.pb, lead 2+ ; oxido dioxo vanadium, $l^ 2-lead 2+ ion divanadiumoylolate
PubChem CID 61471
IUPAC Name lead(2+);oxido(dioxo)vanadium
SMILES [Pb++].[O-][V](=O)=O.[O-][V](=O)=O

Specifications

Physical Form Powder
Quantity 250 g
UN Number UN2291
Merck Index 14,5427
Solubility Information Insoluble in water,decomposes in HNO3
Formula Weight 405.07
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Lead(II) metavanadate
Hazard Category H302+H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
EINECSNumber 233-248-5
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.