Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(Ii) Nitrate, ACS Reagent, Solstice

Catalog No. 6000894 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-008-94 2.5 kg
60-008-93 100 g
60-008-95 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-008-94 Supplier Solstice Supplier No. 2286212.5KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
PubChem CID 24924
ChEBI CHEBI:37187
IUPAC Name lead(2+);dinitrate

Specifications

Quantity 2.5 kg
Linear Formula Pb(NO3)2
UN Number 1469
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Formula Weight 331.21g/mol
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Lead(Ii) Nitrate
Hazard Statement P201-P210-P220-P280-P305 + P351 + P338 + P310-P308 + P313
Precautionary Statement H272-H302 + H332-H318-H360Df-H373-H410
EINECSNumber 233-245-9
RTECSNumber OG2100000
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.