Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(IV) Oxide, ≥97.0%, ACS Reagent, Solstice

Catalog No. 60045460 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-460 100 g Poly Bottle
60-045-461 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-460 Supplier Solstice Supplier No. 237140100G

ACS Reagent,≥97.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.20
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
PubChem CID 14793
IUPAC Name dioxolead
SMILES O=[Pb]=O

Specifications

Color Black
Quantity 100 g
Linear Formula PbO2
UN Number UN1872
Formula Weight 239.2g/mol
Percent Purity ≥97.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Chemical Name or Material Lead(Iv) Oxide
Hazard Statement P201-P210-P220-P280-P308 + P313-P370 + P378
Precautionary Statement H272-H302 + H332-H360Df-H373-H410
DOTInformation 6.1(5.1)/PGIII
EINECSNumber 215-174-5
RTECSNumber OG0700000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.