Learn More
Lead(IV) oxide, 97%
Description
Lead(IV) oxide is used in the manufacture of matches, pyrotechnics, dyes and other chemicals. It is also has several important applications in electrochemistry, in particular in the positive plates of lead acid batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lead(IV) oxide is used in the manufacture of matches, pyrotechnics, dyes and other chemicals. It is also has several important applications in electrochemistry, in particular in the positive plates of lead acid batteries.
Solubility
Insoluble in water. Soluble in dil.HCl.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
| Melting Point | 290°C |
| Quantity | 2500 g |
| UN Number | UN1872 |
| Merck Index | 14,5407 |
| Solubility Information | Insoluble in water. Soluble in dil.HCl. |
| Formula Weight | 239.19 |
| Percent Purity | 97% |
| Odor | Odorless |
| Density | 9.38 |
| Chemical Name or Material | Lead(IV) oxide |
Safety and Handling
| Hazard Category | H272-H302+H332-H350-H360Df-H373 |
| Hazard Statement | GHS H Statement H360-H373-H272-H302-H332 May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. May intensify fire; oxidizer. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: LEAD DIOXIDE |
| EINECSNumber | 215-174-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.