Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(IV) oxide, Puratronic™, 99.995% (metals basis)

Catalog No. AA1072909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1072909 10 g
AA1072918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1072909 Supplier Thermo Scientific Chemicals Supplier No. 01072909
Add to Cart

CAS: 1309-60-0 | O2Pb | 239.20 g/mol

It is employed in manufacture of matches and electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.20
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
PubChem CID 14793
IUPAC Name dioxolead
SMILES O=[Pb]=O

Specifications

Melting Point 290°C (decomposition)
Color Brown to Black
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.995% (Metals basis)
Solubility Information Insoluble in water. Soluble in dilute hydrochloric acid.
Formula Weight 239.19
Odor Odorless
Density 9.38 g/cm3 at 20°C
Chemical Name or Material Lead(IV) oxide
Hazard Category H272-H302+H332-H350-H360Df-H373
Hazard Statement Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 215-174-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.