Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Nitrate, Crystal, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18611241 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-241 25 g Poly Bottle
18-611-236 125 g Poly Bottle
18-611-238 500 g Poly Bottle
18-611-240 2.5 kg Poly Bottle
18-611-237 12 kg Poly Bottle
18-611-239 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-611-241 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L107525GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10099-74-8

Specifications

Physical Form Crystals and/or Chunks
CAS Max % 1
Quantity 25 g
Packaging Poly Bottle
Molecular Weight (g/mol) 331.20
Percent Purity 99%
Grade ACS/Reagent
Chemical Name or Material Lead Nitrate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.