Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Nitrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. L62500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
L62-500 500 g Glass Bottle
L62-100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. L62-500
This item is not returnable. View return policy

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
PubChem CID 24924
ChEBI CHEBI:37187
IUPAC Name λ²-lead(2+) dinitrate
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 470°C
Color White
pH 3 to 4
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99 %
Linear Formula Pb(NO3)2
UN Number 1469
Formula Weight 331.21
Percent Purity ≥99%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure Negligible
Decomposition Information 470°C
Density 4.530g/cm³
Chemical Name or Material Lead Nitrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Suspect cancer hazard. Harmful by inhalation and if swallowed. May cause eye, skin, and respiratory tract irritation. May cause central nervous system effects. May cause harm to the unborn child. Possible risk of impaired fertility. Danger of cumulative effects. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use only under a chemical fume hood. Wear personal protective equipment. Keep away from clothing and other combustible materials. Keep away from clothing and other combustible materials. Use personal protective equipment.

NFPA

Health:2
Flammability:0
Instability:2
DOTInformation DOT Class 5.1, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.