Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Nitrate Stock Solution, 100 ppm Pb, Reagents

Catalog No. 01334834
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-834 1 L
01-334-833 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-834 Supplier Reagents Holdings Llc Supplier No. CL1700001C
Add to Cart
Add to Cart

Lead Nitrate Stock Solution, 100 ppm Pb, Reagent, use for USP lead testing. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Lead Nitrate Stock Solution, 100 ppm Pb
Packaging Glass Bottle
CAS 7732-18-5, 7697-37-2
Specific Gravity 1.01
Physical Form Liquid
Boiling Point 100°C
Synonym Plumbous Nitrate, Lead(II) Nitrate, Lead Dinitrate
Solubility Completely miscible
Quantity 1 L
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.