Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™

Catalog No. SL21500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SL21-500 500 mL Poly Bottle
SL21-100 100 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SL21-500

missing translation for 'validMainframeMsg'

For atomic absorption. (1mL = 1mg Pb) Solute: Lead Metal Solvent: Dilute Nitric Acid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-74-8 , 7697-37-2 , 7732-18-5
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
PubChem CID 24924
ChEBI CHEBI:37187
IUPAC Name λ²-lead(2+) dinitrate
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Poly Bottle
Physical Form Liquid
Concentration 990 to 1010 ppm
Quantity 500 mL
Grade Certified
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Chemical Name or Material Lead, Reference Standard Solution, 1000ppm ±1%
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.