Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead (Shot, Size No. 8), Fisher Chemical

Catalog No. L18500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
L18-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L18-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1/16 in. diameter (1.5875mm)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-92-1 , 7440-36-0 , 7440-38-2
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Color Blue
Melting Point 327.4°C
Boiling Point 1740°C
Atomic Weight 207.19
Density 11.3g/cm³
Quantity 500 g
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 1.3mmHg at 970°C
Form Solid
Formula Weight 207.2
Chemical Name or Material Lead
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Cancer hazard. Harmful by inhalation and if swallowed. May cause skin, eye, and respiratory tract irritation. Possible risk of impaired fertility. Danger of cumulative effects. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Wear personal protective equipment. Ensure adequate ventilation. Use personal protective equipment. Do not get in eyes, on skin, or on clothing. Do not breathe dust. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:0
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.