Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Standard, 1mL = 1mg Pb, 1000ppm Pb, Ricca Chemical
SDP

Catalog No. 430016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
430016 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 430016 Supplier Ricca Chemical Company Supplier No. 430016

Chemical Identifiers

CAS 10099-74-8 , 7697-37-2 , 7732-18-5
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
PubChem CID 24924
ChEBI CHEBI:37187
IUPAC Name λ²-lead(2+) dinitrate
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Poly natural
Concentration or Composition Notes 1000ppm Pb
Equivalence 1 mL = 1 mg Pb
Physical Form Liquid
Color Colorless
Quantity 500 mL
Chemical Name or Material Lead Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.