Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Standard, 5 ppm Pb, Ricca Chemical
SDP

Catalog No. R4298000100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4298000100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R4298000100 Supplier Ricca Chemical Company Supplier No. R4298000100A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 7439-92-1 , 7697-37-2 , 7732-18-5
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Solution
Type Single-component Standard
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Lead Standard, 5 ppm Pb
Show More Show Less
Recommended Storage 15°C - 30°C (59°F - 86°F)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.