Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Subacetate, Spectrum™ Chemical
SDP

Catalog No. 18611258 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-258 500 g Amber Glass Bottle
18-611-256 2.5 kg Amber Glass Bottle
18-611-255 12 kg Poly Pail
18-611-257 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-611-258 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L1088500GM

Chemical Identifiers

CAS 1335-32-6
Molecular Formula C4H10O8Pb3
Molecular Weight (g/mol) 807.70
InChI Key VLOJXAQYHIVPFI-UHFFFAOYSA-H
IUPAC Name tris(λ²-lead(2+)) diacetate tetrahydroxide
SMILES [OH-].[OH-].[OH-].[OH-].[Pb++].[Pb++].[Pb++].CC([O-])=O.CC([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.