Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ LeadCare II Kit
GSA_VA

Catalog No. 23800101 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
48 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-800-101 48 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-800-101 Supplier Meridian Bioscience™ Supplier No. 706762
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Test, educate, and take action, all in one visit using the easiest way to test for lead, LeadCare II. Fingerstick test with a workflow that compliments routine Hgb testing.

  • For in vitro diagnostic use only
  • The test kit is for the quantitative measurement of lead in capillary whole blood
  • This product is for professional use only
  • Sensors and treatment reagent of the test kit are specific for lead only
  • Contents of the LeadCare II Blood Lead Test Kit must only be used with a LeadCare II Analyzer
  • Do not mix components from separate kit lot numbers

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 15°C to 27°C
CE Marker Yes
Detectable Analytes Blood Lead Level
Format Kit
Type Electrochemistry for Blood Lead Level in Capillary Blood
For Use With (Application) LeadCare II Analyzer
Includes 48 sensors, 48 treatment reagent tubes, 50 capillary tubes and plungers, 50 transfer droppers, calibration button, 2 levels of controls
Quantity 48 Tests
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.