Learn More
Learn Your Minerals Kit
Description
Includes
22 specimens: 1.25 x 1.25 in. (3.18 x 3.18cm); streak plate; numbering kit; compartmented collection box; detailed study guide
- Specimens clearly illustrate the properties of hardness, cleavage, fracture, tenacity, and streak
Specifications
Specifications
| Includes | Streak plate; numbering kit; compartmented collection box; detailed study guide |
| Quantity | 22 Specimens |
| Specimen Size | 3.18 x 3.18 cm |