Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Learning To Measure - Building Stem Skills Kit
SDPSCI_ED

Catalog No. S00661 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S00661 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S00661 Supplier United Scientific™ Supplier No. AISMEKIT
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Learning To Measure - Building Stem Skills Kit allows students to take what they learn in the activities and design their own activities.

  • STEM based curriculum kits accent the scientific method and independent inquiry, featuring hands-on learning materials and exciting interactive digital content.
  • Kits contain structured, guided, and open investigations, at introductory, intermediate, and advanced levels, to make inquiry-based learning affordable and fun.
  • Our Learning to Measure Kit contains 5 classroom activities and 5 independent investigations for students to measure lengths and distances, volumes, mass, temperature, and time, and also investigates conservation of mass.
  • The kit includes enough materials for 40 students working in groups of 4, as well as a code to access Teacher and Student Guides and other digital content.

Specifications

Grade 6 to 12
Format Kit
Product Type Measurement Kit
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.