Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lecithin, 60%, egg, Thermo Scientific Chemicals

Catalog No. AAJ6057630
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6057630 250 g
AAJ6057614 25 g
AAJ6057622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6057630 Supplier Thermo Scientific Chemicals Supplier No. J6057630
Add to Cart

missing translation for 'validMainframeMsg'

Lecithin is used as an emulsifying and stabilizing agent in food, pharmaceutical and cosmetic industries. Lecithin is taken as a medicine and is also used in the manufacture of medicines. Soybean lecithin is useful for its application in liquid ram semen. Fractionation process of sunflower lecithin with absolute ethanol was used for obtaining enriched fractions in certain phospholipids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lecithin is used as an emulsifying and stabilizing agent in food, pharmaceutical and cosmetic industries. Lecithin is taken as a medicine and is also used in the manufacture of medicines. Soybean lecithin is useful for its application in liquid ram semen. Fractionation process of sunflower lecithin with absolute ethanol was used for obtaining enriched fractions in certain phospholipids.

Solubility
Soluble in diethyl ether, absolute alcohol, chloroform and petroleum ether. Sparingly soluble in benzene. Insoluble in water and acetone .

Notes
Light, heat, moisture and air-sensitive. Do not store in direct sunlight. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8002-43-5
Molecular Formula C42H80NO8P
Molecular Weight (g/mol) 758.075
MDL Number MFCD00131449
InChI Key JLPULHDHAOZNQI-JLOPVYAASA-N
Synonym 3-sn-phosphatidylcholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine
PubChem CID 16213884
ChEBI CHEBI:86658
IUPAC Name [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC

Specifications

Color Brown to Yellow
Odor Odorless
Quantity 250 g
Beilstein 5209585
Merck Index 14,5428
Solubility Information Soluble in diethyl ether,absolute alcohol,chloroform and petroleum ether. Sparingly soluble in benzene. Insoluble in water and acetone .
Percent Purity 60%
Physical Form Powder
Chemical Name or Material Lecithin, egg
Hazard Category H373
Precautionary Statement P260-P314-P501c
EINECSNumber 232-307-2
RTECSNumber OG7565000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.