Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lecithin, ∽60%, MP Biomedicals™

Catalog No. ICN10214783 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
1 lb.
500 g
5 lb.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10214783 250 g
ICN10214790 500 g
ICN10214801 1 lb.
ICN10214805 5 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10214783 Supplier MP Biomedicals, Inc Supplier No. 0210214783
Add to Cart
Add to Cart

Chemical Identifiers

CAS 8002-43-5
Molecular Formula C42H80NO8P
Molecular Weight (g/mol) 758.075
InChI Key JLPULHDHAOZNQI-JLOPVYAASA-N
Synonym 3-sn-phosphatidylcholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine
PubChem CID 16213884
ChEBI CHEBI:86658
IUPAC Name [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC

Specifications

Color Yellow to Orange
Quantity 250 g
Formula Weight 327.27
Percent Purity ∼60%
Physical Form Solid
Chemical Name or Material Lecithin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.