Learn More
Sigma Aldrich Fine Chemicals Biosciences LB Broth Miller Highly-ref250G
Supplier: Sigma Aldrich Fine Chemicals Biosciences L3522250G
Millers LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids water-soluble vitamins and carbohydrates.