Learn More
United Scientific Supplies LED Array
SDPSCI_ED
Catalog No. S27824 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Power Supply, Activity Guide
Catalog No. Includes
S27824 Power Supply, Activity Guide
1 options
Catalog No. S27824 Supplier United Scientific Supplies Supplier No. LEDA01
Add to Cart
Add to Cart
An easy and dramatic demonstration that depicts the relationship between color and wavelength of light
  • For experiments with color perception, diffraction, and the relationship between frequency and energy
  • Features 10 colored LEDs (spanning from red to violet), and a white LED
  • Display every color at once, or energize one at a time
  • Includes power supply and activity guide
Specifications
Product Type LED Demonstrator
Description 10 Colored LEDs and a White LED
Includes Power Supply, Activity Guide