Learn More
Caron Products And Services LED Plant Growth, 3-Door, 75 cu. ft., 600 mole, 1 Tier
Supplier:  Caron Products And Services 7310752
Encompass_Preferred
LED Plant Growth Chamber. 75 ft3 with 600 mole/m2/s of LED light, 1 tier, temperature control, touchscreen. 10C to 45C, 208/230V, 60 Hz.
Catalog No. NC2676818
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.