Learn More
United Scientific Supplies LED Stroboscope
SDPSCI_ED
Catalog No. S27857 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
7 to 15
Catalog No. Grade
S27857 7 to 15
1 options
Catalog No. S27857 Supplier United Scientific Supplies Supplier No. STROBE01
Add to Cart
Add to Cart
Unique Hand-Held Stroboscope uses high powered LEDs
  • Hand-Held Stroboscope with 30 W Super-LED
  • Delivers bright flashes at rates from 1 Hz to 250 Hz (1 to 15,000 RPM)
  • Frequency adjustable by 2x, 0.5x, or in increments of 0.1 Hz (1 RPM)
  • Magnetic pads on the back facilitate easy attachment to vertical steel surfaces
  • Requires (4) AAA Batteries for three hours of continuous use (not included)
Specifications
Grade 7 to 15
Age 13 to 20
Includes Stroboscope, and Instruction Manual
Requires AAA Batteries (4)
Product Type Stroboscope