Learn More
Ledipasvir acetone, Thermo Scientific™
Specifications
Specifications
| Chemical Name or Material | Ledipasvir acetone |
| CAS | 1441674-54-9 |
| Quantity | 100 mg |
| Shipping Condition | No special requirements |
| Product Line | Life science |
| Molecular Formula | C49 H54 F2 N8 O6 . C3 H6 O |
| Physical Form | Powder |
| Color | White to Yellow |
| Purity | ≥97.5% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.