Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ledipasvir acetone, Thermo Scientific™

Catalog No. AC469365000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469365000 500 mg
AC469361000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC469365000 Supplier Thermo Scientific Chemicals Supplier No. 469365000
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material Ledipasvir acetone
CAS 1441674-54-9
Quantity 500 mg
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C49 H54 F2 N8 O6 . C3 H6 O
Physical Form Powder
Color White to Yellow
Purity ≥97.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.