Learn More
Medchemexpress LLC HY-15602 100mg , Ledipasvir GS-5885 CAS:1256388-51-8 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY15602100MG
Encompass_Preferred
Medchemexpress, HY-15602 100mg Ledipasvir GS-5885 CAS:1256388-51-8 Formula:C49H54F2N8O6 EC50: 34 pM (GT1a), 4 pM (GT1b) Purity:98% Ledipasvir is an inhibitor of the hepatitis C virus NS5A , with EC 50 values of 34 pM against GT1a and 4 pM against GT1b replicon. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2810
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.