Learn More
Abcam Ledol, Antifungal Agent, Solid, Purity >95%, 1 g
Description
- Molecular weight: 222.37 Da
- Purity: >95%
- Physical form: Solid
- Activity: Antifungal agent; putative anti-inflammatory and antinociceptive agent
- Biological effects: May show expectorant, anti-inflammatory, antitussive and antinociceptive effects in vivo
- Administration: Orally active
- Composition: 100% (1aS,4S,4aR,7S,7aR,7bR)-1,1,4,7-tetramethyl-2,3,4a,5,6,7,7a,7b-octahydro-1aH-cyclopropa[e]azulen-4-ol
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Ledol |
| Shipping Condition | Ambient |
| Form | Solid |
| Content And Storage | Store at -20°C. |
| Quantity | 1 g |
| For Use With (Application) | Antifungal, anti-inflammatory, antinociceptive, antitussive, expectorant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.