Learn More
Abcam Ledol, Antifungal Agent, Solid, Purity >95%, 500 mg
Description
- Molecular weight: 222.37 Da
- Purity: >95%
- Form: Solid
- CAS Number: 577-27-5
- Activity: Antifungal, putative anti-inflammatory, and antinociceptive agent
- Additional effects: May show expectorant, anti-inflammatory, antitussive, and antinociceptive effects in vivo
- Administration: Orally active
- Storage buffer: 100% (1aS,4S,4aR,7S,7aR,7bR)-1,1,4,7-tetramethyl-2,3,4a,5,6,7,7a,7b-octahydro-1aH-cyclopropa[e]azulen-4-ol
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Ledol |
| Substrate Properties | Chemical Substrate |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Quantity | 500 mg |
| For Use With (Application) | Antifungal, Anti-inflammatory, Antinociceptive, Expectorant, Antitussive |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.