Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LEF1 Monoclonal Antibody (3A12)
GREENER_CHOICE

Catalog No. 01674334
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-334 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-334 Supplier Invitrogen™ Supplier No. MA112486
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LEF1 Monoclonal Antibody for Western Blot

This gene encodes a transcription factor belonging to a family of proteins that share homology with the high mobility group protein-1. The protein encoded by this gene can bind to a functionally important site in the T-cell receptor-alpha enhancer, thereby conferring maximal enhancer activity. This transcription factor is involved in the Wnt signaling pathway, and it may function in hair cell differentiation and follicle morphogenesis. Mutations in this gene have been found in somatic sebaceous tumors. This gene has also been linked to other cancers, including androgen-independent prostate cancer. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEF1
Applications Western Blot
Classification Monoclonal
Clone 3A12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene LEF1
Gene Accession No. Q9UJU2
Gene Alias 3000002B05; AI451430; DKFZp586H0919; FLJ46390; HGNC:6551; HMG Binding Domain; LEF1; Lef-1; lymphoid enhancer binding factor 1; lymphoid enhancer binding factor 1 short isoform; lymphoid enhancer-binding factor 1; OTTHUMP00000219736; OTTHUMP00000219737; T cell-specific transcription factor 1-alpha; TCF10; TCF1ALPHA; TCF1-alpha; TCF7L3
Gene Symbols LEF1
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues 236-242 of human LEF-1.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51176
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.