Learn More
MSA™ Left/Right™ Ear Muffs
Ear Muffs have dedicated right and left ear cups that are designed to fit ears of all shapes and sizes which offers increased comfort and protection. The headband's breakthrough design simulates the head's curves to provide a comfortable and stable fit, balancing muffs with ear cup angle.
Supplier: MSA™ 10251224
Description
- Precise fit provides maximum loud noise protection using minimal cup size.
- Headband adjusts up and down to provide custom fit for different head sizes.
- Available in low, medium, and high attenuation
Specifications
Yes | |
Yes | |
28 dB | |
Headband Ear Muff | |
No | |
Yes | |
No |
Class C | |
No | |
No | |
Reusable | |
Black | |
Over-the-head | |
EN 352-1 |