Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Legionella Pneumophila Protein, Native, Liquid, 100 μg

Catalog No. 502671857
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-267-1857 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-267-1857 Supplier Abcam Supplier No. AB274665100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Native Legionella pneumophila protein suitable for use in SDS-PAGE and ELISA applications.

  • Target Protein: Legionella pneumophila
  • Form: Liquid
  • Storage buffer: PBS
  • Applications: SDS-PAGE, ELISA
  • Purification: Proprietary technique

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Legionella pneumophila
Substrate Properties Protein-based Substrate
Content And Storage Store at -80°C. Storage buffer: Constituents: PBS.
Label or Dye Unconjugated
Buffer PBS
Form Liquid
Cross Reactivity Legionella pneumophila
Formulation PBS
Quantity 100 μg
For Use With (Application) SDS-PAGE, ELISA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.