Learn More
Leica Microsystems Inoculating Loops and Holders
Description
- Holders constructed of durable, heat-resistant brass with insulating sleeve
- Loops allow an inoculum to be taken from a liquid by using surface tension
Specifications
Specifications
| Material | Brass |
| Length (Metric) | 15.2 cm |
| Length (English) | 6 in. |
| Product Type | Inoculating Loop Holder |