Learn More
Electron Microscopy Sciences EMS Glass Knife Strip Leica Glass 8 mm thick, 25.4 x 400 mm
Supplier: Electron Microscopy Sciences 789008
Specially selected glass strips for making glass knives. Its hardness produces a durable and sharp edge suitable for thick, semi thin, or ultrathin sections.
Thickness: 8 mm
Size: 25.4 x 400 mm