Learn More
Leica Biosystems Leica Paraplast (8-1kg)
Supplier:  Leica Biosystems 39601006/DUP
Encompass_Preferred
Paraplast is recommended for general tissue embedding. It is composed of a refined mixture of highly purified paraffin containing plastic polymers of regulated molecular weights. This supports improved tissue infiltration and quality sections with minimal compression. Paraplast gives brittle, difficult to section tissue an innate elasticity, promoting wrinkle-free sections. 
Catalog No. 50-255-2391
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.