Learn More
THORLABS INC MOIL-10LF, 10 mm Focal Length, Low Magnification Objective Lens
Supplier:  THORLABS INC MOIL10LF
Encompass
The Thorlabs MOIL-10LF is a 10mm focal length, f/1.5 aspheric lens made of N-BK7 glass. It has an anti-reflection coating for the visible spectrum from 350-700nm. The lens has a diameter of 6.35mm and a working distance of 5.0mm. It is designed for laser beam collimation and focusing applications. The lens is optimized for 532nm, but can be used from 350-700nm. It has a damage threshold of 500 MW/cm2 at 532nm. The lens weighs 2 grams.
Catalog No. NC1597569
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.